AC5394386327 Accela ChemBio Inc.

4-(5-Fluoro-2-nitrophenyl)morpholine

  • MDL number MFCD09834357
  • Cas Number : 536977-34-1
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023741&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023741.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart