AC5394386335 Accela ChemBio Inc.

4,4-Dimethyl-2-pentyn-1-amine

  • MDL number MFCD19204088
  • Cas Number : 113439-90-0
  • Molecular Weight : 111.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form Colorless to yellow liquid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023792&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023792.pdf
pictograms No data known
Availability details for
Quantity
  • Availability
  • Add to Cart