AC5394386338 Accela ChemBio Inc.

1-Methyl-4-nitrobenzimidazole

  • MDL number MFCD02181146
  • Cas Number : 31493-66-0
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023819&attname=GoodsCode

Safety Information

signalword Danger
pcodes P261;P280;P301+P312;P302+P352;P305+P351+P338
hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023819.pdf
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart