AC5394386343 Accela ChemBio Inc.

3-Acetamido-2-methylphenyl Acetate

  • MDL number MFCD24335254
  • Cas Number : 76064-16-9
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023845&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023845.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart