AC5394386345 Accela ChemBio Inc.

2,4-Dichloro-5-phenylpyrimidine

  • MDL number MFCD14701757
  • Cas Number : 63558-77-0
  • Molecular Weight : 225.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023852&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023852.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart