AC5394386361 Accela ChemBio Inc.

(S)-3-Aminotetrahydropyran Hydrochloride

  • MDL number MFCD09056862
  • Cas Number : 1245724-46-2
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023980&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY023980.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart