AC5394386378 Accela ChemBio Inc.

2-(5-Methyl-2-thienyl)ethanamine Hydrochloride

  • MDL number MFCD24335263
  • Cas Number : 1215839-34-1
  • Molecular Weight : 177.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024091&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024091.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart