AC5394386379 Accela ChemBio Inc.

3-(Aminomethyl)-5-chloropyridine

  • MDL number MFCD11100748
  • Cas Number : 138402-36-5
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024111&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY024111.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart