AC5394386382 Accela ChemBio Inc.

Ethyl 3-(1,3-Benzodioxol-5-yl)-2-cyanoacrylate

  • MDL number MFCD00022988
  • Cas Number : 2286-56-8
  • Molecular Weight : 245.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024136&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024136.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart