AC5394386383 Accela ChemBio Inc.

Ethyl 3-(2-Bromophenyl)-2-cyanoacrylate

  • MDL number MFCD00175268
  • Cas Number : 59803-32-6
  • Molecular Weight : 280.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024137&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024137.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart