AC5394386392 Accela ChemBio Inc.

5-Amino-4-cyano-1-methylimidazole

  • MDL number MFCD18808604
  • Cas Number : 27700-58-9
  • Molecular Weight : 122.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Light yellow to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024172&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024172.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart