AC5394386394 Accela ChemBio Inc.

N-Boc-3-methylmorpholine-3-carboxylic Acid

  • MDL number MFCD09756545
  • Cas Number : 1052680-53-1
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024196&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024196.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart