AC5394386401 Accela ChemBio Inc.

2-[(Dimethylamino)methyl]benzoic Acid

  • MDL number MFCD08060610
  • Cas Number : 55741-01-0
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024238&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024238.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart