AC5394386404 Accela ChemBio Inc.

4-Amino-5-methylimidazole Dihydrochloride

  • MDL number MFCD18432536
  • Cas Number : 79681-02-0
  • Molecular Weight : 170.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024335&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024335.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart