AC5394386407 Accela ChemBio Inc.

3,5-Difluoro-4-nitrobenzyl Alcohol

  • MDL number MFCD18633079
  • Cas Number : 1123172-89-3
  • Molecular Weight : 189.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024345&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024345.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart