AC5394386418 Accela ChemBio Inc.

4-Amino-1-methylimidazole-5-carbonitrile

  • MDL number MFCD00522606
  • Cas Number : 40637-80-7
  • Molecular Weight : 122.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024466&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024466.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart