AC5394386425 Accela ChemBio Inc.

3-(Boc-amino)pyrrolidin-2-one

  • MDL number MFCD11007813
  • Cas Number : 99780-97-9
  • Molecular Weight : 200.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024478&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024478.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart