AC5394386429 Accela ChemBio Inc.

N-Boc-4-iodo-2-methylaniline

  • MDL number MFCD08703145
  • Cas Number : 666746-27-6
  • Molecular Weight : 333.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024530&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024530.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart