AC5410537500 Accela ChemBio Inc.

2-(Hydroxymethyl)-4-methylmorpholin-3-one

  • MDL number MFCD26383902
  • Cas Number : 1456821-66-1
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024759&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY024759.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart