AC5410537504 Accela ChemBio Inc.

Ethyl (1S,2R)-2-(Cbz-amino)cyclopentanecarboxylate

  • MDL number MFCD26404522
  • Cas Number : 1140972-32-2
  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025349&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025349.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart