AC5410537505 Accela ChemBio Inc.

3-(2-Methoxyethyl)benzylamine

  • MDL number MFCD26383904
  • Cas Number : 1061650-42-7
  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025407&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025407.pdf
pictograms No data known
Availability details for
Quantity
  • Availability
  • Add to Cart