AC5410537507 Accela ChemBio Inc.

2,2-Bis(hydroxymethyl)-4-methylmorpholin-3-one

  • MDL number MFCD26383907
  • Cas Number : 1456821-63-8
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025799&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025799.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart