AC5410537509 Accela ChemBio Inc.

(1S,6R)-3-Boc-7-[(R)-1-phenylethyl]-3,7-diazabicyclo[4.2.0]octane

  • MDL number MFCD26404524
  • Cas Number : 1820574-76-2
  • Molecular Weight : 316.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025942&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025942.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart