AC5410537513 Accela ChemBio Inc.

(2S,4R)-4-Hydroxypyrrolidine-2-carboxamide Hydrochloride

  • MDL number MFCD26404526
  • Cas Number : 32934-42-2
  • Molecular Weight : 166.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026002&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026002.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart