AC5410537516 Accela ChemBio Inc.

5-(Boc-amino)-2-(Boc-aminomethyl)pyridine

  • MDL number MFCD26404527
  • Cas Number : 1456821-59-2
  • Molecular Weight : 323.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026173&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026173.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart