AC5410537517 Accela ChemBio Inc.

5-Bromo-4-methyl-2-nitroanisole

  • MDL number MFCD20482626
  • Cas Number : 1089281-86-6
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Light yellow to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026344&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026344.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart