AC5410537519 Accela ChemBio Inc.

Diisopropyl 5-Methyl-2-thiazoleboronate Lithium Isopropoxide Salt

  • MDL number MFCD26523297
  • Molecular Weight : 293.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026640&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY026640.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart