AC5410537527 Accela ChemBio Inc.

3-(Dimethylamino)-1-(4-methylphenyl)propan-1-one Hydrochloride

  • MDL number MFCD00156711
  • Cas Number : 5250-02-2
  • Molecular Weight : 227.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004002&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004002.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart