AC5410537546 Accela ChemBio Inc.

3'-Bromo-4'-(1-pyrazolyl)acetophenone

  • MDL number MFCD11656810
  • Cas Number : 1186663-58-0
  • Molecular Weight : 265.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY004888&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY004888.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart