AC5410537569 Accela ChemBio Inc.

Ethyl (R)-[(1-Phenylethyl)amino]acetate

  • MDL number MFCD06797321
  • Cas Number : 66512-37-6
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005447&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY005447.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart