AC5410537570 Accela ChemBio Inc.

N-Methyl-2-phenoxybenzylamine

  • MDL number MFCD02089412
  • Cas Number : 361394-74-3
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY005462&attname=GoodsCode

Safety Information

signalword Danger
pcodes P301 + P330 + P331-P280-P305 + P351 + P338-P310-P301+ P312
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY005462.pdf
hstatements H302-H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart