AC5410537589 Accela ChemBio Inc.

6-Amino-1-methylindole

  • MDL number MFCD10000822
  • Cas Number : 135855-62-8
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006491&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY006491.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart