AC5410537619 Accela ChemBio Inc.

2-(Methyl-2-pyridylamino)acetic Acid

  • MDL number MFCD09932969
  • Cas Number : 1016519-61-1
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008636&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008636.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart