AC5410537639 Accela ChemBio Inc.

2-Fluoro-4-methylthio-1-nitrobenzene

  • MDL number MFCD18070982
  • Cas Number : 1025509-81-2
  • Molecular Weight : 187.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010986&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY010986.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart