AC5410537647 Accela ChemBio Inc.

2-(4-Pyridyl)-2-propylamine

  • MDL number MFCD08729286
  • Cas Number : 566155-76-8
  • Molecular Weight : 136.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form Colorless to brown liquid, or white to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011554&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY011554.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart