AC5410537649 Accela ChemBio Inc.

4-Acetyl-3-fluoropyridine

  • MDL number MFCD07375077
  • Cas Number : 87674-21-3
  • Molecular Weight : 139.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011662&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011662.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart