AC5410537650 Accela ChemBio Inc.

1H-Benzotriazole-6-methanamine

  • MDL number MFCD18071016
  • Cas Number : 496841-88-4
  • Molecular Weight : 148.16522

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011665&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY011665.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart