AC5410537655 Accela ChemBio Inc.

(4R,5S)-4-Methyl-5-phenyl-3-propionyl-2-oxazolidinone

  • MDL number MFCD00043400
  • Cas Number : 77877-20-4
  • Molecular Weight : 233.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012056&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012056.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart