AC5410537663 Accela ChemBio Inc.

1-Benzyl-5-(aminomethyl)piperidin-2-one

  • MDL number MFCD18651684
  • Cas Number : 1272756-11-2
  • Molecular Weight : 218.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012354&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012354.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart