AC5410537665 Accela ChemBio Inc.

5-Bromo-N,N-dimethylpyrazole-1-sulfonamide

  • MDL number MFCD18651688
  • Cas Number : 934405-34-2
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012371&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012371.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart