AC5410537666 Accela ChemBio Inc.

N-(2-Propynyl)-2,3-dihydroinden-1-amine

  • MDL number MFCD00033000
  • Cas Number : 1875-50-9
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012373&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012373.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart