AC5410537667 Accela ChemBio Inc.

Ethyl (1R,2S)-2-Aminocyclopentanecarboxylate

  • MDL number MFCD18374515
  • Cas Number : 197916-36-2
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012378&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012378.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart