AC5410537668 Accela ChemBio Inc.

Ethyl (1S,2R)-2-Aminocyclopentanecarboxylate

  • MDL number MFCD18651692
  • Cas Number : 197904-11-3
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012380&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012380.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart