AC5410537670 Accela ChemBio Inc.

4-(1-Methyl-5-imidazolyl)aniline

  • MDL number MFCD18375273
  • Cas Number : 89250-15-7
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012404&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012404.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart