AC5410537673 Accela ChemBio Inc.

Methyl 3-(5-Oxazolyl)benzoate

  • MDL number MFCD06797482
  • Cas Number : 850375-14-3
  • Molecular Weight : 203.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012415&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012415.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart