AC5410537676 Accela ChemBio Inc.

Ethyl 4-(3-Methoxyphenyl)thiazole-2-carboxylate

  • MDL number MFCD06797360
  • Cas Number : 886366-81-0
  • Molecular Weight : 263.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012423&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012423.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart