AC5410537678 Accela ChemBio Inc.

4-(3-Nitrophenyl)thiazole-2-carboxylic Acid

  • MDL number MFCD18375282
  • Cas Number : 185245-05-0
  • Molecular Weight : 250.23064

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012429&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012429.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart