AC5410537684 Accela ChemBio Inc.

Ethyl 3-(5-Oxazolyl)benzoate

  • MDL number MFCD18375301
  • Cas Number : 1261268-84-1
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012501&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012501.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart