AC5410537688 Accela ChemBio Inc.

5-(2-Nitrophenyl)oxazole

  • MDL number MFCD00085146
  • Cas Number : 89808-75-3
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012507&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012507.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart