AC5410537696 Accela ChemBio Inc.

2-Amino-4-(2-bromophenyl)thiazole

  • MDL number MFCD02664013
  • Cas Number : 103965-99-7
  • Molecular Weight : 255.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012620&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012620.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart